Assay Details
| Gene Symbol: Ppp1r14c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133425.3 | NP_596916.1 | 2 - 3 | 993 | 179 | |
| GenBank mRNA | AF407168.3 | - | 2 - 3 | 993 | 179 | |
| BC086978.1 | - | 2 - 3 | 466 | 7191557 | 179 | |
More Information
Set Membership: |
|

Ppp1r14c
Genomic Map