Assay Details
| Gene Symbol: Vsnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012686.2 | NP_036818.2 | 2 - 3 | 390 | 74 | |
| GenBank mRNA | BC092197.1 | - | 2 - 3 | 390 | 7313365 | 74 |
| D10666.1 | - | 2 - 3 | 398 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P534882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812769 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683218 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2769 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177402 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P534882 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P534882 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P534882 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P534882 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P832417 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700327277 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23940 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 25951 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27916 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16710 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 65162 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23382 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11328 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18775 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700327277 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10889218 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889217 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889216 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889215 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889214 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10889213 | - | - |
| Rat Genome 230 2.0 Array | 1368854_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368853_at | - | - |
| Rat Genome 230 2.0 Array | 1368853_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368854_at | - | - |
| Affymetrix HT Rat Focus Array | 1368854_at | - | - |
| Affymetrix HT Rat Focus Array | 1368853_at | - | - |
| Affymetrix HT Rat Focus Array | 1368853_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368854_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600015539.rn.1 | TC0600002133.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600015538.rn.1 | TC0600002133.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368853_at | - | - |
| Rat Expression Set 230 | 1368854_at | - | - |
| Rat Expression Set 230 | 1368854_PM_at | - | - |
| Rat Expression Set 230 | 1368853_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889213 | - | - |
| Rat Genome U34 Set | D10666_at | - | - |
| Rat Genome U34 Set | rc_AI227991_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22425784 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650420 | - | - |
| RatRef-12 Expression BeadChip | ILMN_52723 | - | - |

Vsnl1
Genomic Map