Assay Details
| Gene Symbol: Napsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031670.2 | NP_113858.1 | 2 - 3 | 255 | 64 | |
| GenBank mRNA | AJ251299.1 | - | 2 - 3 | 227 | 64 | |
| BC078790.1 | - | 2 - 3 | 247 | 7109515 | 64 | |
| CB809217.1 | - | 2 - 3 | 281 | 64 | ||
| FQ225885.1 | - | 2 - 3 | 233 | 64 | ||
| FQ230345.1 | - | 2 - 3 | 248 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Napsa
Genomic Map