Assay Details
| Gene Symbol: Pafah1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031763.3 | NP_113951.1 | 896 | 76 | ||
| GenBank mRNA | AF016049.1 | - | 711 | 76 | ||
| BC072510.1 | - | 896 | 7104733 | 76 | ||
| FQ224997.1 | - | 620 | 76 | |||
More Information
Set Membership: |
|

Pafah1b1
Genomic Map