Assay Details
| Gene Symbol: Pak7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107781.1 | NP_001101251.1 | 1 - 2 | 285 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107781.1 transcripts.
Set Membership: |
|

Pak7
Genomic Map