Assay Details
| Gene Symbol: Pak7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107781.1 | NP_001101251.1 | 6 - 7 | 2053 | 103 | |
| XM_008762253.1 | XP_008760475.1 | 3 - 4 | 1556 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pak7
Genomic Map