Assay Details
| Gene Symbol: Pak7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107781.1 | NP_001101251.1 | 7 - 8 | 2189 | 70 | |
| XM_008762253.1 | XP_008760475.1 | 4 - 5 | 1692 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pak7
Genomic Map