Assay Details
| Gene Symbol: Pak7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107781.1 | NP_001101251.1 | 9 - 10 | 2438 | 95 | |
| XM_008762253.1 | XP_008760475.1 | 6 - 7 | 1941 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pak7
Genomic Map