Assay Details
| Gene Symbol: Mios | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173554.1 | NP_001167025.1 | 11 - 12 | 2844 | 66 | |
| XM_006236090.2 | XP_006236152.1 | 10 - 11 | 2844 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mios
Genomic Map