Assay Details
| Gene Symbol: LOC100911453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749373.3 | XP_003749421.1 | 3 - 4 | 648 | 77 | |
| Gene Symbol: Wdr77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008771.3 | NP_001008771.2 | 3 - 4 | 542 | 77 | |
| GenBank mRNA | AY321348.1 | - | 4 - 5 | 485 | 77 | |
| BC097367.1 | - | 3 - 4 | 493 | 7443204 | 77 | |
| FQ213175.1 | - | 3 - 4 | 637 | 77 | ||
| FQ226328.1 | - | 3 - 4 | 537 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911453
Genomic Map