Assay Details
| Gene Symbol: LOC100911453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749373.3 | XP_003749421.1 | 5 - 6 | 764 | 118 | |
| Gene Symbol: Wdr77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008771.3 | NP_001008771.2 | 5 - 6 | 658 | 118 | |
| GenBank mRNA | AY321348.1 | - | 6 - 7 | 601 | 118 | |
| BC097367.1 | - | 5 - 6 | 609 | 7443204 | 118 | |
| FQ213175.1 | - | 5 - 6 | 753 | 118 | ||
| FQ226328.1 | - | 5 - 6 | 653 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC100911453
Genomic Map