Assay Details
| Gene Symbol: Klrb1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085403.1 | NP_001078872.1 | 2 - 3 | 179 | 98 | |
| XM_008763329.1 | XP_008761551.1 | 3 - 4 | 289 | 98 | ||
| XM_008763330.1 | XP_008761552.1 | 3 - 4 | 562 | 98 | ||
| GenBank mRNA | EF100679.1 | - | 2 - 3 | 179 | 98 | |
| EF100685.1 | - | 2 - 3 | 179 | 98 | ||
| X97477.1 | - | 2 - 3 | 179 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klrb1c
Genomic Map