Assay Details
| Gene Symbol: Mrps35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106628.1 | NP_001100098.1 | 3 - 4 | 349 | 72 | |
| XM_006237676.1 | XP_006237738.1 | 3 - 4 | 354 | 72 | ||
| XM_006237677.1 | XP_006237739.1 | 3 - 4 | 360 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrps35
Genomic Map