Assay Details
| Gene Symbol: Gpr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006237520.2 | XP_006237582.1 | 1 - 2 | 2052 | 81 | |
| XM_006237521.2 | XP_006237583.1 | 1 - 2 | 45 | 81 | ||
| XM_006237522.2 | XP_006237584.1 | 1 - 2 | 86 | 81 | ||
| XM_006237523.2 | XP_006237585.1 | 1 - 2 | 45 | 81 | ||
| XM_008763379.1 | XP_008761601.1 | 1 - 2 | 45 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006237521.2, XM_006237522.2, XM_006237523.2, XM_008763379.1 transcripts.
Set Membership: |
|

Gpr19
Genomic Map