Assay Details
| Gene Symbol: E2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750061.3 | XP_003750109.1 | 5 - 6 | 1399 | 74 | |
| XM_008764299.1 | XP_008762521.1 | 5 - 6 | 3012 | 74 | ||
More Information
Set Membership: |
|

E2f2
Genomic Map