Assay Details
| Gene Symbol: Arhgap40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762437.1 | XP_008760659.1 | 2 - 3 | 364 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap40| Gene Symbol: Arhgap40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762437.1 | XP_008760659.1 | 2 - 3 | 364 | 66 | |
Set Membership: |
|