Assay Details
| Gene Symbol: Tcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271277.1 | NP_001258206.1 | 1 - 2 | 133 | 74 | |
| GenBank mRNA | FQ215695.1 | - | 1 - 2 | 131 | 74 | |
| FQ216780.1 | - | 1 - 2 | 133 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tcap
Genomic Map