Assay Details
| Gene Symbol: Fstl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107000.1 | NP_001100470.1 | 5 - 6 | 834 | 56 | |
| XM_008767683.1 | XP_008765905.1 | 2 - 3 | 497 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fstl4
Genomic Map