Assay Details
| Gene Symbol: Amz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014121.1 | NP_001014143.1 | 3 - 4 | 356 | 63 | |
| GenBank mRNA | BC085886.1 | - | 3 - 4 | 356 | 7192275 | 63 |
| FQ212663.1 | - | 3 - 4 | 298 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC085886.1, NM_001014121.1 transcripts.
Set Membership: |
|

Amz2
Genomic Map