Assay Details
| Gene Symbol: Rab11fip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107023.1 | NP_001100493.1 | 11 - 12 | 1405 | 60 | |
| XM_006246903.2 | XP_006246965.1 | 11 - 12 | 1388 | 60 | ||
| XM_006246904.2 | XP_006246966.1 | 9 - 10 | 1350 | 60 | ||
| XM_008768017.1 | XP_008766239.1 | 11 - 12 | 1282 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab11fip4
Genomic Map