Assay Details
| Gene Symbol: Noxo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106986.1 | NP_001100456.1 | 4 - 5 | 712 | 71 | |
| XM_006245948.1 | XP_006246010.1 | 4 - 5 | 337 | 71 | ||
| XM_006245949.1 | XP_006246011.1 | 3 - 4 | 724 | 71 | ||
| XM_006245950.2 | XP_006246012.1 | 3 - 4 | 724 | 71 | ||
| XM_008767566.1 | XP_008765788.1 | 3 - 4 | 724 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Noxo1
Genomic Map