Assay Details
| Gene Symbol: Mettl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108839.1 | NP_001102309.1 | 6 - 7 | 880 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl2b| Gene Symbol: Mettl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108839.1 | NP_001102309.1 | 6 - 7 | 880 | 103 | |
Set Membership: |
|