Assay Details
| Gene Symbol: MGC95210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005532.3 | NP_001005532.2 | 2 - 3 | 652 | 65 | |
| XM_006247676.2 | XP_006247738.1 | 1 - 2 | 283 | 65 | ||
| XM_008768322.1 | XP_008766544.1 | 1 - 2 | 283 | 65 | ||
| XM_008768323.1 | XP_008766545.1 | 1 - 2 | 153 | 65 | ||
| XM_008768324.1 | XP_008766546.1 | 1 - 2 | 155 | 65 | ||
| XM_008768325.1 | XP_008766547.1 | 2 - 3 | 227 | 65 | ||
| XM_008768326.1 | XP_008766548.1 | 2 - 3 | 283 | 65 | ||
| XM_008768328.1 | XP_008766550.1 | 1 - 2 | 283 | 65 | ||
| XM_008768329.1 | XP_008766551.1 | 1 - 2 | 283 | 65 | ||
| GenBank mRNA | BC088445.1 | - | 1 - 2 | 272 | 7126436 | 65 |
| BC099187.1 | - | 2 - 3 | 652 | 7383366 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC088445.1, BC099187.1, NM_001005532.3, XM_006247676.2, XM_008768322.1, XM_008768323.1, XM_008768324.1, XM_008768325.1, XM_008768326.1, XM_008768328.1, XM_008768329.1 transcripts.
Set Membership: |
|

MGC95210
Genomic Map