Assay Details
| Gene Symbol: Tmem88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128155.1 | NP_001121627.1 | 1 - 2 | 218 | 73 | |
| GenBank mRNA | EF532356.1 | - | 1 - 2 | 108 | 73 | |
| EF532357.1 | - | 1 - 2 | 100 | 73 | ||
| EF532358.1 | - | 1 - 2 | 100 | 73 | ||
| EF532359.1 | - | 1 - 2 | 108 | 73 | ||
| EF532360.1 | - | 1 - 2 | 100 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem88
Genomic Map