Assay Details
| Gene Symbol: Myl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106017.1 | NP_001099487.1 | 2 - 3 | 196 | 78 | |
| XM_006251216.2 | XP_006251278.1 | 3 - 4 | 296 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myl7
Genomic Map