Assay Details
| Gene Symbol: Myl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106017.1 | NP_001099487.1 | 4 - 5 | 383 | 58 | |
| XM_006251216.2 | XP_006251278.1 | 5 - 6 | 483 | 58 | ||
| GenBank mRNA | BC161995.1 | - | 383 | 9034230 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Myl7
Genomic Map