Assay Details
| Gene Symbol: Mbtps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC097391.1 | - | 53 | 7453173 | 61 | |
| Gene Symbol: NEWGENE_2324567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773225.1 | XP_008771447.1 | 1 - 2 | 722 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC097391.1, XM_008773225.1 transcripts.
Set Membership: |
|

Mbtps2
Genomic Map