Assay Details
| Gene Symbol: Rtp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107083.1 | NP_001100553.1 | 2 - 3 | 249 | 124 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107083.1 transcripts.
Set Membership: |
|

Rtp2
Genomic Map