Assay Details
| Gene Symbol: Ccar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170472.1 | NP_001163943.1 | 8 - 9 | 807 | 66 | |
| XM_006252268.2 | XP_006252330.1 | 8 - 9 | 924 | 66 | ||
| GenBank mRNA | CA510912.1 | - | 3 - 4 | 199 | 66 | |
| CK596250.1 | - | 5 - 6 | 556 | 66 | ||
| FQ211644.1 | - | 8 - 9 | 918 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccar2
Genomic Map