Assay Details
| Gene Symbol: Gpr174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106938.1 | NP_001100408.1 | 1 - 2 | 80 | 97 | |
| XM_006257163.2 | XP_006257225.1 | 3 - 4 | 301 | 97 | ||
| XM_008773340.1 | XP_008771562.1 | 3 - 4 | 312 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106938.1, XM_006257163.2, XM_008773340.1 transcripts.
Set Membership: |
|

Gpr174
Genomic Map