Assay Details
| Gene Symbol: Mettl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025038.1 | NP_001020209.1 | 4 - 5 | 784 | 72 | |
| GenBank mRNA | BC091261.1 | - | 4 - 5 | 784 | 7377954 | 72 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P017901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6045 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017901 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017901 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P017901 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30547 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104507 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35191 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93388 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33448 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15542 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101962 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 25782 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10830481 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830479 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830483 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830482 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830480 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10830478 | - | - |
| Affymetrix Rat Clariom S Assay | PSR2000014062.rn.1 | TC2000001740.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014063.rn.1 | TC2000001740.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10830478 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21123244 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372931 | - | - |

Mettl24
Genomic Map