Assay Details
| Gene Symbol: LOC102553656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006254368.2 | XP_006254430.1 | 1 - 2 | 308 | 97 | |
| Gene Symbol: Mastl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107369.1 | NP_001100839.1 | 240 | 97 | ||
| XM_006254340.2 | XP_006254402.1 | 302 | 97 | |||
| XM_006254342.2 | XP_006254404.1 | 306 | 97 | |||
More Information
Updated Mapping Information
Set Membership: |
|

LOC102553656
Genomic Map