Assay Details
| Gene Symbol: Fbxo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106206.1 | NP_001099676.1 | 2 - 3 | 876 | 84 | |
| GenBank mRNA | BC158745.1 | - | 2 - 3 | 816 | 9100726 | 84 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fbxo5
Genomic Map