Assay Details
| Gene Symbol: Slc7a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134615.1 | NP_001128087.1 | 6 - 7 | 1418 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc7a14| Gene Symbol: Slc7a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134615.1 | NP_001128087.1 | 6 - 7 | 1418 | 66 | |
Set Membership: |
|