Assay Details
| Gene Symbol: Nceh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127524.2 | NP_001120996.1 | 1 - 2 | 179 | 65 | |
| GenBank mRNA | BC166533.1 | - | 1 - 2 | 179 | 7190480 | 65 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P10081 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P053242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166804 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P10081 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P053242 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P053242 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P10081 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P053242 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P10081 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P191411 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38384 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89961 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10814522 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814526 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814524 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814527 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814525 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814523 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10814521 | - | - |
| Rat Genome 230 2.0 Array | 1374265_at | - | - |
| Rat Genome 230 2.0 Array | 1374265_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200005377.rn.1 | TC0200000889.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200005374.rn.1 | TC0200000889.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10814521 | - | - |
| Rat Genome U34 Set | rc_AI071628_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20893529 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364273 | - | - |

Nceh1
Genomic Map