Assay Details
| Gene Symbol: Ap3m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133593.2 | NP_598277.2 | 7 - 8 | 1036 | 81 | |
| XM_006251629.1 | XP_006251691.1 | 7 - 8 | 1563 | 81 | ||
| XM_006251630.2 | XP_006251692.1 | 7 - 8 | 1082 | 81 | ||
| GenBank mRNA | BC070925.1 | - | 7 - 8 | 1036 | 7096586 | 81 |
| L07073.1 | - | 7 - 8 | 1049 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ap3m1
Genomic Map