Assay Details
| Gene Symbol: Gdf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017203.1 | NP_058899.1 | 1 - 2 | 476 | 80 | |
| GenBank mRNA | AF092733.1 | - | 1 - 2 | 281 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gdf11
Genomic Map