Assay Details
| Gene Symbol: Apof | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024351.1 | NP_001019522.1 | 1 - 2 | 89 | 123 | |
| GenBank mRNA | BC088170.1 | - | 1 - 2 | 89 | 7322324 | 123 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC088170.1, NM_001024351.1 transcripts.
Set Membership: |
|

Apof
Genomic Map