Assay Details
| Gene Symbol: Atp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134364.1 | NP_599191.1 | 2 - 3 | 329 | 72 | |
| GenBank mRNA | BC099743.1 | - | 2 - 3 | 312 | 7937989 | 72 |
| M19044.1 | - | 1 - 2 | 145 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Atp5b
Genomic Map