Assay Details
| Gene Symbol: Gpr34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256675.2 | XP_006256737.1 | 2 - 3 | 2652 | 155 | |
| XM_006256676.2 | XP_006256738.1 | 2 - 3 | 569 | 155 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006256675.2, XM_006256676.2 transcripts.
Set Membership: |
|

Gpr34
Genomic Map