Assay Details
| Gene Symbol: Zer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100707.1 | NP_001094177.1 | 14 - 15 | 2577 | 68 | |
| XM_008761652.1 | XP_008759874.1 | 14 - 15 | 2503 | 68 | ||
| XM_008761653.1 | XP_008759875.1 | 14 - 15 | 2497 | 68 | ||
| XM_008761654.1 | XP_008759876.1 | 14 - 15 | 2488 | 68 | ||
| XM_008761655.1 | XP_008759877.1 | 14 - 15 | 2354 | 68 | ||
| XM_008761656.1 | XP_008759878.1 | 14 - 15 | 2740 | 68 | ||
| XM_008761657.1 | XP_008759879.1 | 14 - 15 | 2689 | 68 | ||
| GenBank mRNA | FQ233933.1 | - | 4 - 5 | 530 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zer1
Genomic Map