Assay Details
| Gene Symbol: Usp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107827.3 | NP_001101297.1 | 10 - 11 | 868 | 57 | |
| XM_006233916.2 | XP_006233978.1 | 10 - 11 | 2003 | 57 | ||
| XM_008761666.1 | XP_008759888.1 | 8 - 9 | 977 | 57 | ||
| XM_008761667.1 | XP_008759889.1 | 8 - 9 | 935 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp20
Genomic Map