Assay Details
| Gene Symbol: Tbx19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107193.1 | NP_001100663.1 | 4 - 5 | 871 | 58 | |
| XM_008769651.1 | XP_008767873.1 | 5 - 6 | 1609 | 58 | ||
| XM_008769652.1 | XP_008767874.1 | 6 - 7 | 970 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbx19
Genomic Map