Assay Details
| Gene Symbol: Tbx19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107193.1 | NP_001100663.1 | 5 - 6 | 1051 | 65 | |
| XM_008769651.1 | XP_008767873.1 | 6 - 7 | 1789 | 65 | ||
| XM_008769652.1 | XP_008767874.1 | 7 - 8 | 1150 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbx19
Genomic Map