Assay Details
| Gene Symbol: Fam163a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109072.1 | NP_001102542.1 | 4 - 5 | 521 | 67 | |
| XM_008769668.1 | XP_008767890.1 | 4 - 5 | 760 | 67 | ||
| XM_008769669.1 | XP_008767891.1 | 4 - 5 | 757 | 67 | ||
| XM_008769670.1 | XP_008767892.1 | 4 - 5 | 757 | 67 | ||
| XM_008769671.1 | XP_008767893.1 | 4 - 5 | 601 | 67 | ||
| XM_008769672.1 | XP_008767894.1 | 2 - 3 | 438 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P137257 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P137262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275596 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157945 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P137257 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P137262 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P137262 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P137257 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P137262 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P137257 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55980 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52240 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10768932 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768933 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10768931 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300011765.rn.1 | TC1300001690.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300011771.rn.1 | TC1300001690.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300011764.rn.1 | TC1300001690.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300011767.rn.1 | TC1300001690.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300011770.rn.1 | TC1300001690.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10768931 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21463638 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350161 | - | - |

Fam163a
Genomic Map