Assay Details
| Gene Symbol: Cdc42ep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230806.2 | XP_006230868.1 | 2 - 3 | 310 | 86 | |
| XM_008760142.1 | XP_008758364.1 | 1 - 2 | 898 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006230806.2, XM_008760142.1 transcripts.
Set Membership: |
|

Cdc42ep2
Genomic Map