Assay Details
| Gene Symbol: LOC102550148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765369.1 | XP_008763591.1 | 1 - 2 | 314 | 88 | |
| Gene Symbol: Otogl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765370.1 | XP_008763592.1 | 40 - 41 | 7122 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC102550148
Genomic Map