Assay Details
| Gene Symbol: Rit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013060.1 | NP_001013078.1 | 3 - 4 | 546 | 72 | |
| GenBank mRNA | BC091382.1 | - | 3 - 4 | 546 | 7320994 | 72 |
| FQ213009.1 | - | 3 - 4 | 343 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rit2
Genomic Map