Assay Details
| Gene Symbol: Slc1a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108973.1 | NP_001102443.1 | 2 - 3 | 454 | 58 | |
| XM_008763892.1 | XP_008762114.1 | 2 - 3 | 383 | 58 | ||
| GenBank mRNA | JF422064.1 | - | 2 - 3 | 256 | 58 | |
| JF422066.1 | - | 2 - 3 | 256 | 58 | ||
| JF422067.1 | - | 2 - 3 | 256 | 58 | ||
| JF422068.1 | - | 2 - 3 | 256 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a7
Genomic Map