Assay Details
| Gene Symbol: Ccnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108110.1 | NP_001101580.1 | 1 - 2 | 159 | 88 | |
| XM_008765745.1 | XP_008763967.1 | 1 - 2 | 319 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnt1
Genomic Map